Learn More
FMR1 Mouse anti-Drosophila, Clone: 6A15, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051512
Catalog No. 05-151-2MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-FMR1 Antibody, clone 6A15 is an antibody against FMR1 for use in western blotting.
Specifications
Specifications
FMR1
6A15
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Purified
Murine
IgG1, κ, κ
100μg
Primary
Drosophila
Western Blot
Unconjugated
NP_611645.1
FMR1, FMRP, FRAXA, POF, POF1
His-tagged recombinant protein corresponding to Drosophila FMR1.
Protein G purfied
−20°C for up to 12 months.
Monoclonal