Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNBP1 Rabbit anti-Equine, Human, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523414

Catalog No. PIPA523414


Add to cart

Description

Description

FNBP1 is a regulatory protein belonging to the formin-binding-protein family. It has a FCH domain, a coiled-coil domain, a Rho family protein-binding domain (RBD), and a SH3 domain. FNBP1 interacts with sorting nexin 2 (SNX2), tankyrase (TNKS), and dynamin and is an important regulator of endocytosis. It self associates and binds to lipids to promote membrane invagination and the formation of tubules. It acts as a link between RND2 signaling and regulation of the actin cytoskeleton. FNBP1 induces neurite branching in response to ARHN. The MLL-FNBP1 fusion protein with MLL has very low transforming ability. FNBP1 is associated with acute myelogemous leukemia. FNBP1 is very highly expressed in epithelial cells, brown adipose tissue, cardiomyocytes, enteric ganglia and glucagon-producing cells of the pancreas, germ cells of the testis and brain.
Specifications

Specifications

FNBP1
Polyclonal
Unconjugated
FNBP1
Liquid
FNBP1
Synthetic peptide (597 DRESPDGSYTEEQSQESE 614) of FNBP1 protein.
Protein A
RUO
Antibody
Polyclonal
Equine, Human, Mouse, Non-human primate, Rat
Immunohistochemistry (Paraffin)
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q96RU3, Q80TY0, Q8R511
FBP17, formin-binding protein 1, formin-binding protein 17
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100069871, 14269, 192348, 23048
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit