Learn More
farnesyltransferase, CAAX box, alpha, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human FNTA protein.
Supplier: Abnova Corporation H00002339B01
Description
Prenyltransferases attach either a farnesyl group or a geranylgeranyl group in thioether linkage to the cysteine residue of protein's with a C-terminal CAAX box. CAAX geranylgeranyltransferase and CAAX farnesyltransferase are heterodimers that share the same alpha subunit but have different beta subunits. This gene encodes the alpha subunit of these transferases. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MAATEGVGEAAQGGEPGQPAQPPPQPHPPPPQQQHKEEMAAEAGEAVASPMDDGFVSLDSPSYVLYRDRAEWADIDPVPQNDGPNPVVQIIYSDKFRDVYDYFRAVLQRDERSERAFKLTRDAIELNAANYTVWHFRRVLLKSLQKDLHEEMNYITAIIEEQPKNYQVWHHRRVLVEWLRDPSQELEFIADILNQDAKNYHAWQHRQWVIQEFKLWDNELQYVDQLLKEDVRNNSVWNQRYFVISNTTGYNDRAVLEREVQYTLEMIKLVPHNESAWNYLKGILQDRGLSKYPNLLNQLLDLQPSHSSPYLIAFLVDIYEDMLENQCDNKEDILNKALELCEILAKEKDTIRKEYWRYIGRSLQSKHSTENDSPTNVQQSpecifications
farnesyltransferase, CAAX box, alpha | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human FNTA protein. | |
FNTA | |
FPTA/MGC99680/PGGT1A/PTAR2 | |
Mouse | |
50 μL | |
Signal Transduction | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
BC017029 | |
FNTA | |
FNTA (AAH17029, 1 a.a. ∼ 379 a.a) full-length human protein. | |
RUO | |
Yes | |
2339 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.