Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FNTA, Polyclonal, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232180

Catalog No. NBP232180

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

FNTA
1.0 mg/mL
Western Blot 1:2000 - 1:10000, Immunohistochemistry, Immunoprecipitation 2 - 10 ug/mg lysate
NP_002018.1
CAAX farnesyltransferase subunit alpha, EC 2.5.1.58, EC 2.5.1.59, farnesyl-protein transferase alpha-subunit, farnesyltransferase, CAAX box, alpha, FPTA, FTase-alpha, GGTase-I-alpha, MGC99680, PGGT1A, protein farnesyltransferase/geranylgeranyltransferase type-1 subunit alpha, protein prenyltransferase alpha subunit repeat containing 2, PTAR2, Ras proteins prenyltransferase subunit alpha, type I protein geranyl-geranyltransferase alpha subunit, Type I protein geranyl-geranyltransferase subunit alpha
Rabbit
IgG
100 ul
Signal Transduction
Primary
2339
Western Blot, Immunohistochemistry, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
FNTA
The immunogen this antibody was made to, maps to a region between residue 329 to 379 of human Farnesyltransferase, CAAX Box, Alpha using the numbering given in entry NP_002018.1 (GeneID 2339).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only