Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Follicle Stimulating Hormone, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA185445

Catalog No. PA185445

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

FSH is a hormone that regulates the development, growth, pubertal maturation, and reproductive processes of the human body.

PA1-85445 has exhibited the following cross reactivities: Human FSH - 66%, Rat LH - 8%, Human LH beta - 6%, Human CG alpha - 6%, Rat TSH - 4%. No reactivity was observed with human GH, prolactin, ACTH or mouse GH.
Specifications

Specifications

Follicle Stimulating Hormone
Polyclonal
whole serum with 0.09% sodium azide
P01225, Q60687
FSHB
Murine FSH.
50μL
Store at 4°C or Store at -20°C if preferred
Primary
2488
Immunohistochemistry (Frozen), Western Blot
Unconjugated
FSHB
Serum
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit