Learn More
Follistatin Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA186870
Catalog No. PIPA186870
Add to cart
Description
Description
Reconstitute with 0.1 mL of distilled water. Prior to reconstitution, store at 4°C. After reconstitution, store undiluted at -20°C, avoiding freeze/thaw cycles. Rabbit anti Human Follistatin antibody recognizes human follistatin (FST), a single-chain glycosylated secreted protein of gonadal origin, originally identified as an antagonist of activin and suppressor of follicle stimulating hormone (FSH) synthesis/secretion.
Follistatin is a single-chain gonadal protein that specifically inhibits follicle-stimulating hormone release. The single FST gene encodes two isoforms, FST317 and FST344 containing 317 and 344 amino acids respectively, resulting from alternative splicing of the precursor mRNA. In a study in which 37 candidate genes were tested for linkage and association with polycystic ovary syndrome (PCOS) or hyperandrogenemia in 150 families, evidence was found for linkage between PCOS and follistatin.
Specifications
Specifications
Follistatin
Polyclonal
Unconjugated
FST
Lyophilized
FST
Recombit human follistatin
Affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
1 mg/mL
PBS with no preservative; pH 7.2
P19883
FS, Follictatin
Rabbit
IgG
100 μg
4° C
Primary
10468