Learn More
Fos Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06341
Catalog No. 06-341-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect Fos with Anti-Fos Antibody (Rabbit Polyclonal Antibody), that has been shown to work in immunoprecipitation, western blot, immunocytochemistry.
Specifications
Specifications
Fos
Unconjugated
Affinity Purified
Rabbit
IgG
100μg
Primary
NM_005252
Immunocytochemistry, Immunoprecipitation, Western Blot
PBS containing 2mg/ml BSA and 0.2% sodium azide
FOS; G0S7; c-fos
Trp E fusion-protein corresponding to the N-terminal fragment containing amino acid residues 75-155 of human v-Fos
Immunoaffinity chromatography
−20°C for 12 months
Polyclonal
Human, Murine