Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fra2 Rabbit anti-Hamster, Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524452

Catalog No. PIPA524452

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with chicken, mouse and rat based on sequence homology.

The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation.
Specifications

Specifications

Fra2
Polyclonal
PBS with 0.09% sodium azide
P15408
FRA2, FRA-2, fos-related antigen 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100755919, 2355
Immunohistochemistry, Western Blot
Unconjugated
FOSL2
Liquid
FOSL2
KLH conjugated synthetic peptide between 95-122 amino acids from the central region of human FOSL2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit