Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXA1 Mouse anti-Human, Mouse, Clone: 2F83, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051466
Description
Use Anti-FOXA1 Antibody, clone 2F83 (Mouse Monoclonal Antibody) validated in western blot, immunocytochemistry, immunohistochemistry (paraffin) to detect FOXA1 also known as Forkhead box protein A1, hepatocyte nuclear factor 3 alpha.Specifications
FOXA1 | |
Monoclonal | |
Unconjugated | |
NP_937987 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2F83 | |
Purified mouse monoclonal antibody in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
HNF-3-alpha, HNF-3A, HNF3A, TCF3A | |
Recombinant human GST-FOXA1 protein encompassing amino acids 7-86. | |
100 μg | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only