Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-FoxC1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224834
Description
FoxC1 Polyclonal specifically detects FoxC1 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FoxC1 | |
Polyclonal | |
Western Blot 1:250-1: 1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin | |
Q12948 | |
FOXC1 | |
A portion of amino acids 250-300 of human FOXC1 was used as the immunogen. | |
0.2 mL | |
Cancer | |
2296 | |
Human, Primate | |
Antisera |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Whole antisera with 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
ARA, FKHL7RIEG3, forkhead box C1, forkhead box protein C1, forkhead, drosophila, homolog-like 7, forkhead/winged helix-like transcription factor 7, forkhead-related activator 3, Forkhead-related protein FKHL7, Forkhead-related transcription factor 3, FREAC-3, FREAC3IRID1, IGDA, IHG1, myeloid factor-delta | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a) and chimpanzee, and 88% homologous in monkey. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only