Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXC1 Rabbit anti-Human, Non-human primate, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523160
Description
FOXC1 is a protein belonging to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this protein is not known, however, it plays a role in the regulation of FGF19-FGFR4-MAPK pathway to promote both the development and maintece of anterior segment structures within the eye. Mutations in this gene cause various glaucoma, iridogoniodysgenesis anomaly, Peters anomaly that includes central corneal leukoma, absence of the posterior corneal stroma and Descemetmembrane and Axenfeld-Rieger anomaly characterized by posterior corneal embryotoxon, iris adhesion to the Schwalbe line, hypertelorism, hypoplasia of the malar bones, congenital absence of some teeth and mental retardation.Specifications
FOXC1 | |
Polyclonal | |
FOXC1 | |
ARA, FKHL7, FREAC-3, FREAC3, IGDA, IHG1, IRID1, RIEG3, forkhead box protein C1, forkhead, drosophila, homolog-like 7, forkhead-related activator 3, forkhead-related protein FKHL7, forkhead-related transcription factor 3, forkhead/winged helix-like transcription factor 7, myeloid factor-delta | |
Rabbit | |
200 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Q12948 | |
FOXC1 | |
Synthetic peptide (279 SARPLSLDGADSAP 292) of FOXC1 protein. | |
RUO | |
2296 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction