Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXF2 Monoclonal Antibody (3D11), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA518898
Description
Peptide Sequence: SPYLKQPPAL TPSSNPAASA GLHSSMSSYS LEQSYLHQNA REDLSVGLPR YQHHSTPVCD RKDFVLNFNG ISSFHPSASG SYYHHHHQSV CQDIKPCV
FOXF2 Monoclonal antibody specifically detects FOXF2 in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
FOXF2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q12947 | |
FOXF2 | |
FOXF2 (NP_001443, 346 a.a. ∽ 443 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Immunocytochemistry, Western Blot | |
3D11 | |
Unconjugated | |
FOXF2 | |
Fkh20; FKHL6; forkhead box F2; forkhead box protein F2; forkhead homolog 20; forkhead-like 6; forkhead-related activator 2; Forkhead-related protein FKHL6; forkhead-related transcription factor 2; FOXF2; FREAC2; FREAC-2; LOW QUALITY PROTEIN: forkhead box protein F2; LUN; Protein LUN | |
Mouse | |
Protein A | |
RUO | |
2295 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction