Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXG1, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131654

Catalog No. PA131654

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon.

PA1-31654 detects FOXG1 from human samples. Expected to cross react with rat and mouse due to sequence homology.PA1-31654 has been successfully used in Western blot applications. A suggested positive control for this product is human brain lysate.The PA1-31654 immunogen is: Peptide with sequence HQNQGSSSNPLIH, from C Terminus of the protein sequence according to NP_004462 (FOXG1A); NP_005240 (FOXG1B)
Specifications

Specifications

FOXG1
Polyclonal
Unconjugated
FOXG1
Purified
FOXG1
Peptide with sequence HQNQGSSSNPLIH, from C Terminus of the protein sequence according to NP_004462 (FOXG1A); NP_005240 (FOXG1B)
IgG fraction
RUO
Antibody
Polyclonal
Human, Murine, Rat
Western Blot
0.5mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P55316, Q60987, Q00939
BF1, BF2, FHKL3, FKH2, FKHL1, FKHL2, FKHL3, FKHL4, FOXG1A, FOXG1B, FOXG1C, HBF-1, HBF-2, HBF-3, HBF-G2, HBF2, HFK1, HFK2, HFK3, KHL2, QIN
Goat
IgG
50μg
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
2290
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit