Learn More
FOXL1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human FOXL1 peptide using ARM Technology.
Manufacturer:  Abnova Corporation H00002300K
Catalog No. 89-200-457
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Description
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line
Order Info
1. Customer may provide cell or tissue lysate for antibody screening.
2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
Specifications
FOXL1
1mg
forkhead box L1
FOXL1
Liquid
FOXL1
A synthetic peptide of human FOXL1
1mg x up to 3 Abs
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Western Blot
Unconjugated
In 1X PBS, pH 7.4
FOXL1
FKH6, FKHL11, FREAC7
Rabbit
IgG
RUO
Primary
2300