Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXL2 Goat anti-Bovine, Human, Mouse, Rat, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131950

Catalog No. PIPA131950


Add to cart

Description

Description

PA1-31950 detects FOXL2 from human samples. PA1-31950 has been successfully used in Western blot applications. The PA1-31950 immunogen is: Synthetic peptide: DSKTGALHSRLDL, corresponding to amino acids 364-376 of FOX L2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

FOXL2 is a winged helix/forkhead transcription factor that is highly conserved in human, goat, mouse, and certain aquatic species.  Given its cross-species conservation, it appears to play a critical role evolutionarily, likely in ovarian differentiation
Specifications

Specifications

FOXL2
Polyclonal
Unconjugated
FOXL2
Liquid
FOXL2
synthetic peptide DSKTGALHSRLDL, corresponding to amino acids 364-376 of FOX L2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Human, Mouse, Rat
ELISA, Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q6VFT7, P58012, O88470
forkhead box L2, forkhead transcription factor FOXL2
Goat
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
26927, 281770, 367152, 668
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit