Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXL2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523675

Catalog No. PIPA523675

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a forkhead transcription factor. The protein contains a fork-head DNA-binding domain and may play a role in ovarian development and function. Mutations in this gene are a cause of blepharophimosis syndrome and premature ovarian failure 3.
Specifications

Specifications

FOXL2
Polyclonal
PBS with 0.09% sodium azide
P58012
BPES, BPES1, PFRK, PINTO, POF3, forkhead box protein L2, forkhead transcription factor FOXL2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
668
Flow Cytometry, Western Blot
Unconjugated
FOXL2
Liquid
FOXL2
KLH conjugated synthetic peptide between 3-29 amino acids from the N-terminal region of human FOXL2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit