Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXO1 Mouse anti-Human, Mouse, Rat, Clone: 2H8.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051075
Description
Use Anti-FOXO1 Antibody, clone 2H8.2 (Mouse Monoclonal Antibody) validated in western blot, immunocytochemistry, immunohistochemistry (paraffin) to detect FOXO1 also known as Forkhead box protein O1A, Forkhead in rhabdomyosarcoma.Specifications
FOXO1 | |
Monoclonal | |
Unconjugated | |
NP_002006.2 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2H8.2 | |
Purified mouse monoclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
FKH1, FKHR, FOXO1A, OTTHUMP00000018301 | |
GST-tagged recombinant protein corresponding to full length FOXO1. | |
200 μg | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only