Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXO1 Rabbit anti-Bovine, Canine, Human, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523132

Catalog No. PIPA523132


Add to cart

Description

Description

FOXO1 (FKHR, ForKHead Box 01) is a member in a subfamily of the forkhead homeotic gene family of transcription factors. Recent experiments have shown that FOXO1 can act as either a coactivator or a corepressor of nuclear receptor activity that is mediated through the LXXLL motif found in the carboxyl terminal region of the FKHR protein. Association of FOXO1 with PAX3 has been implicated in alveolar rhabdomyosarcoma. Recent studies link the anti-tumor activity of FOXO1, the process of autophagy and myogenic growth and differentiation. FOXO1 is the main target of insulin signaling that regulated the metabolic homeostasis in response to oxidative stress. FOXO1 binds to the insulin response element (IRE) with consensus sequence 5 ft.-TT[G/A]TTTTG-3 ft. and the related Daf-16 family binding element (DBE) with consensus sequence 5 ft.- TT[G/A]TTTAC-3 ft.. FOXO1 is a regulator of redox balance, osteoblast numbers and controls bone mass. Further, FOXO1 orchestrates the endocrine function of the skeleton in regulating glucose metabolism, and suppresses the transcriptional activity of RUNX2, an upstream activator of osteocalcin/BGLAP. In hepatocytes, FOXO1 promotes gluconeogenesis by acting together with PPARGC1A to activate the expression of genes such as IGFBP1, G6PC and PPCK1. FOXO1 is an important regulator of cell death acting downstream of CDK1, PKB/AKT1 and SKT4/MST1.
Specifications

Specifications

FOXO1
Polyclonal
Unconjugated
FOXO1
Liquid
FOXO1
Synthetic peptide (251 RRRAASMDNNSKFAK 265) of FOXO1A protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Canine, Human, Mouse, Non-human primate, Rat
Western Blot
1.0 mg/mL
PBS with 0.05% sodium azide
E1BPQ1, Q12778, Q9R1E0, G3V7R4
FKH1, FKHR, FOXO1A, forkhead box protein O1, forkhead box protein O1A, forkhead, Drosophila, homolog of, in rhabdomyosarcoma
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2308, 477295, 506618, 56458, 84482
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit