Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXO4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA520975

Catalog No. PIPA520975


Add to cart

Description

Description

A suggested positive control is A-20 cell lysate. PA5-20975 can be used with blocking peptide PEP-1089.

FOXO4 is a ubiquitously expressed protein member of a subfamily of the forkhead homeotic gene family of transcription factors and shuttles between the cytoplasm and nucleus. FOXO transcription factors are key players of cell fate decisions, metabolism, stress resistance, tumor suppression and are regulated by growth factors, oxidative stress or nutrient deprivation. In the absence of PI3K/AKT activation, FOXO4 localizes in the nucleus where it functions as a transcription factor. FOXO4 can also be phosphorylated by JNK following induction of reactive oxygen species (ROS), resulting in transcriptional activation and the induction of a negative feedback mechanism to counteract the ROS. It is through this mechanism that FOXO4 is thought to sensitize cancer cells to doxorubicin-mediated toxicity.
Specifications

Specifications

FOXO4
Polyclonal
Unconjugated
FOXO4
Liquid
FOXO4
A 15 amino acid peptide near the amino terminus of human FOXO4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P98177, Q9WVH3
AFX, AFX1, MLLT7; fork head domain transcription factor AFX1; forkhead box protein O4; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 7
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
302415, 4303, 54601
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit