Learn More
FoxP1 Mouse, Unlabeled, Clone: JC12, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 564213
Description
The JC12 monoclonal antibody specifically binds to Forkhead box protein P1 (FoxP1). FoxP1 belongs to subfamily P of the forkhead box (FOX) winged-helix transcription factor family. It plays a variety of roles such as regulating the formation of lymphoid, lung, brain and heart tissues. FoxP1 is critical in regulating early B cell development and is also required for maintenance of naοve T cell quiescence, and monocyte differentiation and macrophage function. Aberrant expression of FOXP1 has been linked with mucosa-associated lymphoid tissue lymphoma and diffuse-large B cell lymphoma.
Specifications
FoxP1 | |
Monoclonal | |
0.5mg/mL | |
Forkhead box protein P1; hFKH1B; HSPC215; QRF1; 12CC4 | |
do not include immunogen on TDS | |
50 μg | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
JC12 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Human, Mouse | |
IgG2a κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.