Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP1 Mouse, PE, Clone: JC12, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564216

Catalog No. BDB564216


Add to cart

Description

Description

The JC12 monoclonal antibody specifically binds to Forkhead box protein P1 (FoxP1). FoxP1 belongs to subfamily P of the forkhead box (FOX) winged-helix transcription factor family. It plays a variety of roles such as regulating the formation of lymphoid, lung, brain and heart tissues. FoxP1 is critical in regulating early B cell development and is also required for maintenance of naοve T cell quiescence, and monocyte differentiation and macrophage function. Aberrant expression of FOXP1 has been linked with mucosa-associated lymphoid tissue lymphoma and diffuse-large B cell lymphoma.

Specifications

Specifications

FoxP1
JC12
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG2a, κ
RUO
Primary
Human, Murine
Flow Cytometry
0.2mg/mL
Forkhead box protein P1; hFKH1B; HSPC215; QRF1; 12CC4
Affinity Purified
do not include immunogen on TDS
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit