Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FoxP2, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP151907

Catalog No. NBP151907


Add to cart

Description

Description

FoxP2 Polyclonal antibody specifically detects FoxP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications

Specifications

FoxP2
0.5 mg/mL
Western Blot 0.5 - 2 ug/mL, Peptide ELISA 1:128000
NP_055306.1
CAG repeat protein 44, CAGH44TNRC10forkhead box protein P2, DKFZp686H1726, forkhead box P2, forkhead/winged-helix transcription factor, SPCH1, trinucleotide repeat containing 10, Trinucleotide repeat-containing gene 10 protein
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognise all three reported isoforms (NP_055306.1; NP_683696.2; NP_683697.1).
ELISA, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
FOXP2
Peptide with sequence C-REIEEEPLSEDLE corresponding to C-Terminus according to NP_055306.1, NP_683696.2, NP_683697.1.
Immunogen affinity purified
RUO
Primary
93986
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only