Learn More
FOXP3 Rabbit anti-Human, Mouse, DyLight® 488, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522948
Catalog No. PIPA522948
Add to cart
Description
Description
This antibody reacts weakly with human samples.
FOXP3 (Forkhead box protein 3) is a member of the forkhead/winged-helix family of transcriptional regulators, highly conserved across mammals, and essential for normal immune homeostasis. FOXP3 is 381 amino acids long, stably and constitutively expressed at a high level in CD25 + CD4 positive regulatory T cells, a low level in CD4-positive/CD25-negative cells, and is absent in CD4-negative/CD8-positive T cells. FOXP3 may be a master regulatory gene, and a more specific marker of regulatory T cells. Defects in the gene encoding FOXP3 protein cause the scurfy phenotype in mice. In humans FOXP3 defects play a role in IPEX syndrome (immune dysfunction, polyendocrinopathy, enteropathy, X-linked syndrome), also known as X-linked autoimmunity-allergic dysregulation (XLAAD) syndrome. Transcript variants of FOXP3 encoding different isoforms have been identified. In human breast and colon cancer cells, expression of FOXP3 is regulated by p53 in response to the DNA damage.
Specifications
Specifications
FOXP3
Polyclonal
DyLight™ 488
FOXP3
Liquid
FOXP3
A synthetic peptide made to the N-terminal region of human FOXP3 (between residues 1-50).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1 mg/mL
sodium borate with no preservative
Q9BZS1, Q99JB6
JM2, AIID, DIETER, IPEX, PIDX, XPID, FOXP3delta7, forkhead box protein P3, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, immunodeficiency, polyendocrinopathy, enteropathy, X-linked, scurfin
Rabbit
IgG
100 μL
4° C, store in dark
Primary
20371, 50943