Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXP3 Rabbit anti-Human, Mouse, DyLight® 550, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522776

Catalog No. PIPA522776


Add to cart

Description

Description

This antibody has shown reactivity against Ms (CD4+CD25+ T cells), does not react with Hu Jurkat, Ms CD4+CD25- T cell lysates.

FOXP3 (Forkhead box protein 3) is a member of the forkhead/winged-helix family of transcriptional regulators, highly conserved across mammals, and essential for normal immune homeostasis. FOXP3 is 381 amino acids long, stably and constitutively expressed at a high level in CD25 + CD4 positive regulatory T cells, a low level in CD4-positive/CD25-negative cells, and is absent in CD4-negative/CD8-positive T cells. FOXP3 may be a master regulatory gene, and a more specific marker of regulatory T cells. Defects in the gene encoding FOXP3 protein cause the scurfy phenotype in mice. In humans FOXP3 defects play a role in IPEX syndrome (immune dysfunction, polyendocrinopathy, enteropathy, X-linked syndrome), also known as X-linked autoimmunity-allergic dysregulation (XLAAD) syndrome. Transcript variants of FOXP3 encoding different isoforms have been identified. In human breast and colon cancer cells, expression of FOXP3 is regulated by p53 in response to the DNA damage.
Specifications

Specifications

FOXP3
Polyclonal
DyLight™ 550
FOXP3
Liquid
FOXP3
A peptide correpsonding to amino acids 43-100 of mouse FOXP3
Protein G
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Western Blot
1.15 mg/mL
sodium borate with 0.05% sodium azide
Q9BZS1, Q99JB6
RP23-54C14.1, JM2, scurfin, sf, forkhead box protein P3, scurfy
Rabbit
IgG
100 μL
4° C, store in dark
Primary
20371, 50943
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit