Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP3 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $337.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide corresponding to the exon 2 deletion site was used as immunogen.
Antigen FoxP3
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224397SS
View Documents
Novus Biologicals
NBP224397SS
0.025 mg
Each for $119.60
Add to cart
 
NBP224397
View Documents
Novus Biologicals
NBP224397
0.1 mg
Each for $337.00
Add to cart
 
Description

Description

FoxP3 Polyclonal antibody specifically detects FoxP3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications

Specifications

Rabbit
A synthetic peptide corresponding to the exon 2 deletion site was used as immunogen.
Polyclonal
Primary
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Q9BZS1
PBS with 0.05% Sodium Azide
IgG
FoxP3
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b, CRA_c, and CRA_d), opossum, chimpanzee, rhesus monkey, horse, dog, mouse, cow and rat, 78% homologous in zebrafish and 71% homologous in chicken. In human, the four isoforms range in size from 941 amino acids to 1008 amino acids, with theoretical molecular weights of approximately 103kD to 110kD.
50943
1.0 mg/ml
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit