Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FPR2 Mouse anti-Human, Clone: GM1D6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA117769

Catalog No. 01-670-182


Add to cart

Description

Description

This antibody recognizes native human FPRL1 protein expressed in transfected mammalian cells.

Human formyl peptide-receptor-like-1 (FPRL1) belongs to the large family of G-protein coupled receptors (GPCR). It is a seven transmembrane protein expressed on mononuclear phagocytes and microglial cells. FPRL1 is a member of the chemoattractant subfamily of G protein-coupled receptors and plays a key role in inflammation via chemotaxis and the regulation of mediator release from leukocytes. It interacts with formyl peptides to attract phagocytes to sites of infection and promote inflammatory reactions. FPRL1 also interacts with amyloid beta peptides and has been implicated in phagocyte attraction to sites of amyloid plaques in Alzheimer’s disease. Since FPRL1 is expressed in neutrophils and monocytes, and it was shown using another monoclonal antibody that chemokines can be potent and specific ligands, FPRL1 might have interesting functions in inflammatory pathways.
Specifications

Specifications

FPR2
Monoclonal
2 mg/mL
PBS with no preservative; pH 7.2
P25090
Formyl Peptide Receptor Like-1, FPRH1, HM63, ALXR, LXA4R, fMLPR1
Mouse
IgG1, kappa
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2358
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence
GM1D6
Unconjugated
FPR2
Liquid
FPR2
cDNA encoding human FPRL1
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit