Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FPR3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA132544

Catalog No. PIPA132544


Add to cart

Description

Description

PA1-32544 detects FPRL2 from human samples. PA1-32544 has been successfully used in immunohistochemistry (paraffin tissue) applications. The PA1-32544 immunogen is: Synthetic peptide, KLH conjugated.

Specifications

Specifications

FPR3
Polyclonal
Unconjugated
FPR3
Liquid
FPR3
Synthetic peptide, KLH conjugated.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.7
P25089
FML2_HUMAN, FMLPY, FPRH1, FPRH2, FPRL2, RMLP-R-I
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2359
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit