Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fra2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA129171

Catalog No. PIPA129171

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-29177 detects MADD from human samples. PA1-29177 has been successfully used in Western blot applications. The PA1-29177 immunogen is: Synthetic peptide corresponding to the second half of human MADD protein.

Cellular oncogenes, or proto-oncogenes, play pivotal roles in cellular communication pathways that regulate normal growth, development and differentiation. The cellular oncogene families fos and jun encode nuclear proteins that can function as transcripti
Specifications

Specifications

Fra2
Polyclonal
Unconjugated
FOSL2
Liquid
FOSL2
Synthetic peptide corresponding to the C-terminus of human Fra2 protein.
Protein A
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.2% BSA and 0.1% sodium azide; pH 7.4
P15408
Fos-Related Antigen-2, FosL2
Rabbit
IgG
500 μL
4° C, do not freeze
Primary
2355
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit