Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Frizzled-3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130940

Catalog No. NBP130940


Add to cart

Description

Description

Frizzled-3 Polyclonal antibody specifically detects Frizzled-3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Frizzled-3
1.0mg/mL
Western Blot, Immunohistochemistry 1:10 - 1:500
Q9NPG1
frizzled (Drosophila) homolog 3, frizzled homolog 3 (Drosophila), frizzled-3, Fz-3, hFz3
Rabbit
IgG
Antigen Affinity-purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Antibody reactive against peptide, detects immunogen. Further validation in progress.
Immunohistochemistry, Western Blot
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Affinity Purified
FZD3
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Frizzled 3. The exact sequence is proprietary.
76 kDa
100 ul
GPCR, Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway
Primary
7976
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only