Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FZD7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA131056

Catalog No. PA131056


Add to cart

Description

Description

PA1-31056 detects Frizzled 7 from human samples. PA1-31056 has been successfully used in immunohistochemistry (paraffin tissue) applications. The PA1-31056 immunogen is: Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive) Store at 4°C short term. For extended storage aliquot and store at -20°C or below. Avoid freeze-thaw cycles.

A member of the frizzled (fz) family, which has been shown to be the transmembrane receptors for the Wnt molecules. Wnts have been implicated in embryonic induction, cell fate determination, control of cell polarity and maligt transformation.
Specifications

Specifications

FZD7
Polyclonal
Unconjugated
FZD7
Liquid
FZD7
Synthetic peptide with a proprietary sequence.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.7
O75084
FZD7
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
8324
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit