Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FRMPD1, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136957

Catalog No. NBP136957


Add to cart

Description

Description

FRMPD1 Polyclonal antibody specifically detects FRMPD1 in Human, Canine samples. It is validated for Peptide ELISA.
Specifications

Specifications

FRMPD1
0.5 mg/mL
Peptide ELISA Detection limit 1:2000
NP_055722.1
FERM and PDZ domain containing 1, FERM and PDZ domain-containing protein 1, FRMD2KIAA0967FERM domain-containing protein 2, MGC138833
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody recognizes and epitope within the internal region.
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
FRMPD1
Peptide with sequence C-PQPETEEEDRDLEAH corresponding to internal region according to NP_055722.1.
Immunogen affinity purified
RUO
Primary
22844
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only