Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFKM Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515437

Catalog No. PIPA515437


Add to cart

Description

Description

Phosphofructokinase catalyzes the irreversible conversion of fructose 6 phosphate to fructose 1,6 bisphosphate. Mammalian PFK is a complex isozyme consisting of 3 subunits: muscle (M), liver (L), and platelet (P). Only M type PFK isozyme is expressed in mature muscle, while erythrocytes contain both L and M subunits. Defects in PFKM are the cause of glycogen storage disease type 7 (GSD7), also known as Tarui disease.
Specifications

Specifications

PFKM
Polyclonal
PBS with 0.09% sodium azide
P08237
6-phosphofructokinase, muscle type; Phosphofructokinase 1; Phosphohexokinase; Phosphofructo-1-kinase isozyme A; PFK-A; Phosphofructokinase-M; PFKX
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5213
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
PFKM
Liquid
PFKM
KLH conjugated synthetic peptide between 746-776 amino acids from the C-terminal region of human Fructose 6 Phosphate Kinase (PFKM)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit