Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FUBP3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP150044

Catalog No. NBP150044


Add to cart

Description

Description

FUBP3 Polyclonal antibody specifically detects FUBP3 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

FUBP3
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 mcg/mg
Affinity Purified
FUBP3
The immunogen for this product maps to a region between residue 1 and 50 of human FUBP3 using the numbering given in entry NP_003925.1 (GeneID 8939).
Immunogen affinity purified
RUO
Primary
8939
Immunoprecipitation, Western Blot
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
far upstream element (FUSE) binding protein 3, far upstream element-binding protein 3, FBP3, FLJ25229, FUSE-binding protein 3
Rabbit
IgG
0.1 ml
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only