Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523696
Description
This antibody is predicted to react with bovine based on sequence homology.
FUS Polyclonal antibody specifically detects FUS in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
FUS | |
Polyclonal | |
Unconjugated | |
FUS | |
16) in malignant liposarcoma); 16) malignant liposarcoma; 75 kDa DNA-pairing protein; ALS6; D430004D17Rik; D930039C12Rik; ETM4; FUS; FUS RNA binding protein; Fus1; fused in sarcoma; fused in sarcoma RNA binding protein; fusion (involved in t(12; fusion gene in myxoid liposarcoma; fusion, derived from t(12; fusion, derived from t(12;16) malignant liposarcoma; fus-like protein; heterogeneous nuclear ribonucleoprotein P2; hnRNP P2; HNRNPP2; oncogene FUS; Oncogene TLS; pigpen protein; POMP75; Protein pigpen; RNA-binding protein FUS; TLS; translocated in liposarcoma; translocated in liposarcoma protein | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
233908, 2521 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P35637, P56959 | |
FUS | |
KLH conjugated synthetic peptide between 499-526 amino acids from the C-terminal region of human FUS | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction