Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FUS2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130978

Catalog No. NBP130978


Add to cart

Description

Description

FUS2 Polyclonal antibody specifically detects FUS2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

FUS2
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
NAA80
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human FUS2. The exact sequence is proprietary.
31 kDa
100 ul
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Pig (85%). Not yet tested in any other species.
Western Blot
Unconjugated
PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal
FUS-2, FUS2EC 2.3.1.-, N-acetyltransferase 6, N-acetyltransferase 6 (GCN5-related), Protein fus-2, Protein fusion-2
Rabbit
IgG
Antigen Affinity-purified
RUO
Primary
24142
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only