Learn More
FUT6 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA524850
Catalog No. PIPA524850
Add to cart
Description
Description
FUT6 is a Golgi stack membrane protein that is involved in the creation of sialyl-Lewis X, an E-selectin ligand.
Specifications
Specifications
FUT6
Polyclonal
Unconjugated
FUT6
Liquid
FUT6
KLH conjugated synthetic peptide between 102-130 amino acids from the central region of human FUT6
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P51993
FCT3A, FT1A, Fuc-TVI, FucT-VI, alpha-(1,3)-fucosyltransferase, fucosyltransferase VI, galactoside 3-L-fucosyltransferase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2528