Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FYB Mouse anti-Human, Unlabeled, Clone: 5, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610945

 View more versions of this product

Catalog No. BDB610945

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Engagement of the T cell receptor (TcR) results in the activation of multiple intracellular src protein tyrosine kinases (PTKs) including p56 [lck] and p59 [fyn]. PTK activation induces TcRζ phosphorylation and the subsequent recruitment and activation of ZAP-70 which specifically phosphorylates SLP-76. SLP-76 interacts with multiple components of T cell signaling pathways, including Grb-2, PLC-γ, Vav and SLAP-130 (also known as SLP-76 Associated Phosphoprotein of 130kDa). SLAP-130 contains multiple tyrosine based motifs, proline base type I and II SH3 domain binding motifs, lysine/glutamic acid rich nuclear localization motifs, and an SH3-like domain. It associates with the SH2 domain of SLP-76 and is phosphorylated by TcR induced PTKs. This protein has also been reported to directly interact with fyn and has been referred to as fyn binding protein (FYB). Co-transfection data has shown that a limited concentration of FYB/SLAP-130 enhances TcR-induced IL-2 production, while overexpression downregulates gene expression. Thus, FYB/SLAP-130 is viewed to be a regulatory component of the fyn/SLP-76 signaling cascade in T cells.

Immunofluorescence, Western Blotting

Specifications

Specifications

FYB
5
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Immunofluorescence, Western Blot
250μg/mL
Fyn Binding Protein; SLAP-130
Affinity Purified
Human FYB aa. 673-783
150μg
Cell Biology
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.