Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FYCO1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP147265

Catalog No. NBP147265


Add to cart

Description

Description

FYCO1 Polyclonal antibody specifically detects FYCO1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

FYCO1
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 5-15 ug/mg lysate
Affinity Purified
FYCO1
The immunogen for this product maps to a region between residue 325 and 375 of human FYVE and Coiled-Coil Domain Containing 1 using the numbering given in entry NP_078789.2 (GeneID 79443).
Immunogen affinity purified
RUO
Primary
79443
Immunoprecipitation, Western Blot
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
DKFZp779K1152, FLJ13335, FYVE and coiled-coil domain containing 1, FYVE and coiled-coil domain-containing protein 1, MGC126517, RUFY3, RUN and FYVE domain containing 3, ZFYVE7MGC126519, Zinc finger FYVE domain-containing protein 7
Rabbit
IgG
0.1 ml
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only