Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fyn Mouse, Unlabeled, Clone: 25, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610164

 View more versions of this product

Catalog No. BDB610164


Add to cart

Description

Description

Stimulation of the T cell antigen receptor (TcR) results in protein tyrosine phosphorylation via non-receptor tyrosine kinases. The src-related p59[fyn] kinase is one such enzyme that associates with the TcR. Ligation of the TcR activates the protein kinase activity of p59[fyn] invarious human T cells. Fyn interacts with the CD3-ζ chains through its N-terminal region. In turn, fyn binds other proteins through its SH2 and SH3 domains. These proteins (p82 and p116) may serve as substrates and/or mediators of fyn activity. A 72kDa fyn-related protein binds to the TcR/CD3 complex in certain T cells. This protein, known as p72[fyn-R], is a variant of the fyn kinase and contains at least four novel Ser/Thr phosphorylation sites. These sites may make p72[fyn-R] less susceptible to the action of phosphatases than p59[fyn]. Like p59[fyn], p72[fyn-R] is constitutively associated with its receptor. The exact relationship between these two proteins is not yet clear. However, p72[fyn-R] immunoprecipitates with various antisera raised to either the N- or C-terminus of fyn. Similar to src and yes, fyn is highly
expressed in brain. The localization of fyn in mouse brain and its interaction with myelin-associated glycoprotein (MAG) suggests that the fyn kinase has a role in the sensory network and in myelination during early stages of CNS formation.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Fyn
25
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA
150μg
Cell Biology
Primary
Canine, Human, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.