Learn More
Fyn Mouse, Unlabeled, Clone: 25, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610163
Description
Stimulation of the T cell antigen receptor (TcR) results in protein tyrosine phosphorylation via non-receptor tyrosine kinases. The src-related p59[fyn] kinase is one such enzyme that associates with the TcR. Ligation of the TcR activates the protein kinase activity of p59[fyn] invarious human T cells. Fyn interacts with the CD3-ζ chains through its N-terminal region. In turn, fyn binds other proteins through its SH2 and SH3 domains. These proteins (p82 and p116) may serve as substrates and/or mediators of fyn activity. A 72kDa fyn-related protein binds to the TcR/CD3 complex in certain T cells. This protein, known as p72[fyn-R], is a variant of the fyn kinase and contains at least four novel Ser/Thr phosphorylation sites. These sites may make p72[fyn-R] less susceptible to the action of phosphatases than p59[fyn]. Like p59[fyn], p72[fyn-R] is constitutively associated with its receptor. The exact relationship between these two proteins is not yet clear. However, p72[fyn-R] immunoprecipitates with various antisera raised to either the N- or C-terminus of fyn. Similar to src and yes, fyn is highly
expressed in brain. The localization of fyn in mouse brain and its interaction with myelin-associated glycoprotein (MAG) suggests that the fyn kinase has a role in the sensory network and in myelination during early stages of CNS formation.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Fyn | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
50 μg | |
Cell Biology | |
Canine, Human, Rat | |
IgG2b |
Western Blot | |
25 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.