Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fyn (pY528)/c-Src (pY530) Mouse, Unlabeled, Clone: 31, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612668

Catalog No. BDB612668


Add to cart

Description

Description

p59fyn is a member of the Src family of protein tyrosine kinases. Two isoforms of the protein have been identified, designated as FynB and Fyn T. Fyn B has been shown to be localized to the brain, whereas Fyn T associates with both B and T cells. Stimulation of the T cell antigen receptor (TcR) results in protein tyrosine phosphorylation via non-receptor tyrosine kinases. p59fyn kinase (Fyn T) associates with the TcR. Ligation of the TcR activates the protein kinase activity of p59fyn in various human T cells. Fyn interacts with the CD3-zeta chains through its N-terminal region. In turn, fyn binds other proteins through its SH2 and SH3 domains. These proteins (p82 and p116) may serve as substrates and/or mediators of fyn activity. p59fyn kinase is tyrosine phosphorylated at two sites, Tyr-417 (autophosphorylation site), and Tyr-528 (negative regulatory site).

Intracellular Staining, Western Blotting

Specifications

Specifications

Fyn (pY528)/c-Src (pY530)
31
Unconjugated
Affinity Purified
Phosphorylated Human Fyn (Y528) Peptide
50μg
Cell Biology
Primary
Human, Murine
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit