Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FZD4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526481

Catalog No. PIPA526481

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with chicken, mouse and rat based on sequence homology.

FZD4 may play a role as a positive regulator of the Wingless type MMTV integration site signaling pathway.
Specifications

Specifications

FZD4
Polyclonal
PBS with 0.09% sodium azide
Q9ULV1, Q61088
CD344, EVR1, FEVR, FZD4S, Fz-4, FzE4, GPCR, WNT receptor frizzled-4, frizzled 4, seven transmembrane spanning receptor, frizzled homolog 4, frizzled-4, hFz4
Rabbit
Ig
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
14366, 8322
Western Blot
Unconjugated
FZD4
Liquid
FZD4
KLH conjugated synthetic peptide between 495-522 amino acids from the C-terminal region of human FZD4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit