Learn More
Seracare Life Sciences Inc ANTI-G PIG IGG (H+L) AB FITC
Supplier: Seracare Life Sciences Inc 52300303
Affinity purified polyclonal antibody to Guinea Pig IgG, both heavy and light chains (whole IgG) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
This item is not returnable.
View return policy