Learn More
GΒ Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06238
Catalog No. 06-238-MI
Add to cart
Description
Description
Detect Gβ using this Anti-Gβ Antibody validated for use in immunoprecipitation and western blotting.
Specifications
Specifications
Unconjugated
Serum
Rabbit
IgG
50μL
Primary
NM_002074
Immunoprecipitation, Western Blot
PBS, pH 7.2
GNB2; SG; IG
Internal sequence of G-protein β subunit (KTREGNVRVSREL) conjugated to octavalent polylysine base
Antiserum
−20°C for 24 months
Polyclonal
Human, Murine
Documents
Promotions
Promotions
  • MilliporeSigma
    Buy two qualifying MilliporeSigma antibodies and get a third of equal or lesser value at no additional cost.
    01-31-2021