Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNAI2, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527496

Catalog No. PIPA527496

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is an alpha subunit of guanine nucleotide binding proteins (G proteins). The encoded protein contains the guanine nucleotide binding site and is involved in the hormonal regulation of adenylate cyclase. Several transcript variants encoding different isoforms have been detected for this gene, but the full-length nature of only two are known so far.

PA5-27496 targets G protein alpha Inhibitor 2 in WB applications and shows reactivity with Human and mouse samples.The PA5-27496 immunogen is recombinant fragment corresponding to a region within amino acids 1 and 235 of Human G protein alpha Inhibitor 2.
Specifications

Specifications

G protein alpha Inhibitor 2
Polyclonal
Unconjugated
GNAI2
Purified
GNAI2
Recombinant fragment corresponding to a region within amino acids 1 and 235 of Human G protein alpha Inhibitor 2
Antigen affinity chromatography
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
2771
Western Blot
0.39mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P04899, P08752
GIP, GNAI2B, H_LUCA15.1, H_LUCA16.1, GTP-binding regulatory protein Gi alpha-2 chain, adenylate cyclase-inhibiting G alpha protein, guanine nucleotide-binding protein G(i) subunit alpha-2, guanine nucleotide-binding protein G(i), alpha-2 subunit
Rabbit
IgG
100μL
Antibody
Polyclonal
Human, Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit