Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNAI2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527520

Catalog No. PIPA527520


Add to cart

Description

Description

PA5-27520 targets G protein alpha Inhibitor 2 in IF and WB applications and shows reactivity with Human samples. The PA5-27520 immunogen is recombit fragment corresponding to a region within amino acids 94 and 355 of Human G protein alpha Inhibitor 2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is an alpha subunit of guanine nucleotide binding proteins (G proteins). The encoded protein contains the guanine nucleotide binding site and is involved in the hormonal regulation of adenylate cyclase. Several transcript variants encoding different isoforms have been detected for this gene, but the full-length nature of only two are known so far.
Specifications

Specifications

GNAI2
Polyclonal
Unconjugated
GNAI2
Liquid
GNAI2
Recombit fragment corresponding to a region within amino acids 94 and 355 of Human G protein alpha Inhibitor 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
P04899
GIP, GNAI2B, H_LUCA15.1, H_LUCA16.1, GTP-binding regulatory protein Gi alpha-2 chain, adenylate cyclase-inhibiting G alpha protein, guanine nucleotide-binding protein G(i) subunit alpha-2, guanine nucleotide-binding protein G(i), alpha-2 subunit
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2771
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit