Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBL Rabbit anti-Bovine, Canine, Chicken, Human, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141327

Catalog No. 01-675-664


Add to cart

Description

Description

This antibody is 93% homologous to xenopus.

Principal Names: G-protein beta subunit like (GBL protein); MGC111011 Official Gene Symbol: GBL Gen Bank Accession Number: NP_071767 Gene ID: 64223 (Human) Gene Map Locus: 16p13.3 (Human) LST8, a 36-40KDa intracellular protein, is a novel member of WD-40 protein family. It is a highly conserved cytosolic protein consisting of seven WD-40 repeating motifs. LST8 is a functional component of mTOR signaling complex (TORC1) and positively up-regulates the mTOR pathway. It interacts with the kinase domain of mTOR and stabilizes its interaction with raptor. LST8 participates in regulating cell growth and also in nutrient and growth factor-mediated mTOR signaling to S6K1. Reports suggest up-regulated expression of LST8 due to insulin indicating a possible role of LST8 in intracellular events related to insulin-receptor activation including insulin-stimulated glucose transport and glycogen synthesis. Northern Blot analysis detected a ubiquitous expression of LST8 with highest expression in brain and testis.
Specifications

Specifications

GBL
Polyclonal
Unconjugated
MLST8
Liquid
MLST8
Synthetic peptide corresponding to residues 150-200 of human LST8.
Protein G
RUO
Antibody
Polyclonal
Bovine, Canine, Chicken, Human, Mouse, Non-human primate, Rat
Western Blot
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
Q17QU5, Q9BVC4, Q9DCJ1, Q9Z2K5
GBL, LST8
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
416558, 535236, 56716, 610936, 64223, 64226
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit