Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR116 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA132512

Catalog No. PIPA132512


Add to cart

Description

Description

PA1-32512 detects GPCR GPR116 from human samples. PA1-32512 has been successfully used in immunohistochemistry (paraffin tissue) applications. The PA1-32512 immunogen is: Synthetic peptide, KLH conjugated.

Specifications

Specifications

GPR116
Polyclonal
Unconjugated
ADGRF5
Liquid
ADGRF5
Synthetic peptide, KLH conjugated.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.7
Q8IZF2
DKFZp564O1923, FLJ90640, KIAA0758, KPG_001
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
221395
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit