Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR119, Polyclonal, Invitrogen™

Catalog No. PIPA120479
Change view
Click to view available options
Quantity:
25 μg
Catalog No. Quantity
PIPA120479 25 μg
1 options

Catalog No. PIPA120479

Supplier: Thermo Scientific PA120479

Rabbit Polyclonal Antibody

GPCR GPR119 is a member of the G protein coupled receptor (GPCR) family. Members of this protein family contain seven putative transmembrane domains and may mediate signaling processes to the interior of the cell via activation of heterotrimeric G proteins.

PA1-20479 detects G Protein-Coupled Receptor GPR119 in human samples.PA1-20479 has been successfully used in immunohistochemistry (FFPE) procedures.PA1-20479 immunogen corresponds to a synthetic peptide located in the 3rd cytoplasmic domain (aa sequence is considered to be commercially sensitive).
TRUSTED_SUSTAINABILITY

Specifications

Antigen G Protein-Coupled Rec. 119
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene GPR119
Gene Accession No. Q8TDV5
Gene Alias RP1-20I3.4, GPCR2, MGC119957
Gene Symbols GPR119
Host Species Rabbit
Immunogen Synthetic peptide located in the 3rd cytoplasmic loop of G Protein-Coupled Receptor GPR119
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139760
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.