Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA130767

Catalog No. PIPA130767

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

PA1-30767 detects GPR15 from rat, mouse, human samples. PA1-30767 has been successfully used in Western blot applications. The PA1-30767 immunogen is: Synthetic Peptide - amino acids 13-28 of human GPR15. The sequence differs from those of African green monkey and pig-tailed macaque BOB by one amino acid Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

GPR15
Polyclonal
Unconjugated
P49685, Q0VDU3
GPR15
Synthetic Peptide corresponding to amino acids 13-28 of human GPR15. The sequence differs from those of African green monkey and pig-tailed macaque BOB by one amino acid
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot
0.5 mg/mL
GPR15
MGC126828, MGC126830
Rabbit
Antigen affinity chromatography
RUO
2838, 288181, 71223
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit